Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UGLYAH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_117809.6 | NP_193438.2 | 6 - 7 | 599 | 74 | |
GenBank mRNA | AK118019.1 | - | 6 - 7 | 484 | 74 | |
GQ303359.1 | - | 6 - 7 | 438 | 74 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |