Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KAN3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_117878.3 | NP_567535.1 | 2 - 3 | 868 | 89 | |
GenBank mRNA | AY048690.1 | - | 2 - 3 | 647 | 89 |
Set Membership: |
Amplicon length between 86 and 100 Recommended Assay Arabidopsis thaliana Probe spans exons |