Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IP5PII | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001341251.1 | NP_001319979.1 | 2 - 3 | 471 | 142 | |
NM_117911.4 | NP_567547.1 | 2 - 3 | 562 | 142 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |