Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DEG5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001341278.1 | NP_001328913.1 | 2 - 3 | 687 | 123 | |
NM_001341279.1 | NP_001328914.1 | 2 - 3 | 622 | 123 | ||
NM_117947.5 | NP_567552.2 | 2 - 3 | 702 | 123 | ||
GenBank mRNA | AK176772.1 | - | 2 - 3 | 472 | 123 | |
AY056227.1 | - | 2 - 3 | 461 | 123 | ||
AY091427.1 | - | 2 - 3 | 418 | 123 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |