Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DEG5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_117947.5 | NP_567552.2 | 6 - 7 | 1078 | 122 | |
GenBank mRNA | AK176772.1 | - | 6 - 7 | 848 | 122 | |
AY056227.1 | - | 6 - 7 | 837 | 122 | ||
AY091427.1 | - | 6 - 7 | 794 | 122 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |