Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IRX1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_117994.4 | NP_567564.1 | 5 - 6 | 922 | 138 | |
GenBank mRNA | AY099636.1 | - | 5 - 6 | 822 | 138 | |
KJ676982.1 | - | 5 - 6 | 755 | 138 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |