Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NRAMP5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_117995.2 | NP_193614.1 | 3 - 4 | 1442 | 101 | |
GenBank mRNA | AJ292076.1 | - | 3 - 4 | 1366 | 101 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |