Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RFC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001343715.1 | NP_001330213.1 | 19 - 20 | 2525 | 94 | |
NM_147883.6 | NP_680188.1 | 19 - 20 | 2525 | 94 | ||
GenBank mRNA | BT014965.1 | - | 17 - 18 | 2323 | 94 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |