Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RFC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_147883.6 | NP_680188.1 | 21 - 22 | 2868 | 120 | |
GenBank mRNA | BT014965.1 | - | 19 - 20 | 2666 | 120 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |