Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GTB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001084306.2 | NP_001077775.1 | 8 - 9 | 969 | 62 | |
NM_001198388.1 | NP_001185317.1 | 9 - 10 | 907 | 62 | ||
NM_001334206.1 | NP_001321759.1 | 8 - 9 | 1042 | 62 | ||
NM_105218.5 | NP_176723.3 | 8 - 9 | 988 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |