Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KOR2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_105234.3 | NP_176738.1 | 4 - 5 | 1706 | 117 | |
GenBank mRNA | AK229402.1 | - | 4 - 5 | 1563 | 117 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |