Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SMP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_105239.6 | NP_564859.2 | 2 - 3 | 317 | 99 | |
GenBank mRNA | BT002797.1 | - | 2 - 3 | 248 | 99 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |