Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CLPP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_105338.4 | NP_564880.1 | 3 - 4 | 795 | 120 | |
GenBank mRNA | AB022328.1 | - | 3 - 4 | 594 | 120 | |
AY087349.1 | - | 3 - 4 | 647 | 120 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |