Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WRKY9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_105485.4 | NP_176982.1 | 1 - 2 | 302 | 98 | |
GenBank mRNA | AB493525.1 | - | 1 - 2 | 281 | 98 | |
AY052645.1 | - | 1 - 2 | 281 | 98 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |