Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARG1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_105508.4 | NP_177004.1 | 4 - 5 | 704 | 130 | |
GenBank mRNA | AF089810.1 | - | 4 - 5 | 612 | 130 | |
BT025556.1 | - | 4 - 5 | 451 | 130 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |