Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WRKY57 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001334406.1 | NP_001321562.1 | 3 - 4 | 1110 | 134 | |
NM_105598.3 | NP_177090.1 | 3 - 4 | 1341 | 134 | ||
NM_202383.3 | NP_974112.1 | 3 - 4 | 1341 | 134 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Arabidopsis thaliana Probe spans exons |