Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SRO3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001334450.1 | NP_001321500.1 | 2 - 3 | 816 | 137 | |
NM_105712.3 | NP_177201.1 | 2 - 3 | 816 | 137 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |