Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MMZ2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001198436.1 | NP_001185365.1 | 3 - 4 | 480 | 75 | |
NM_105734.3 | NP_564994.1 | 3 - 4 | 450 | 75 | ||
GenBank mRNA | BT004011.1 | - | 3 - 4 | 435 | 75 | |
BT004943.1 | - | 3 - 4 | 290 | 75 |
Gene Symbol: AT1G70650 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001198435.1 | NP_001185364.1 | 8 - 9 | 1681 | 75 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |