Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NdhL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_105744.4 | NP_177233.1 | 2 - 3 | 285 | 72 | |
GenBank mRNA | BT010638.1 | - | 2 - 3 | 220 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |