Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HB18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_105760.6 | NP_177248.3 | 1 - 2 | 306 | 83 | |
GenBank mRNA | AK118149.1 | - | 1 - 2 | 163 | 83 | |
AK175837.1 | - | 1 - 2 | 163 | 83 | ||
AK175965.1 | - | 1 - 2 | 163 | 83 | ||
BT005594.1 | - | 1 - 2 | 74 | 83 | ||
EU339186.1 | - | 1 - 2 | 244 | 83 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |