Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SOL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_105827.3 | NP_177314.2 | 12 - 13 | 1475 | 76 | |
NM_202397.2 | NP_974126.1 | 13 - 14 | 1333 | 76 | ||
GenBank mRNA | AJ555410.1 | - | 13 - 14 | 1212 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |