Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ALAAT2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001084342.1 | NP_001077811.1 | 4 - 5 | 533 | 62 | |
NM_001198451.1 | NP_001185380.1 | 4 - 5 | 543 | 62 | ||
NM_105892.5 | NP_565040.2 | 4 - 5 | 600 | 62 | ||
GenBank mRNA | AF275371.1 | - | 4 - 5 | 514 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |