Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SUS5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001344166.1 | NP_001318683.1 | 7 - 8 | 1378 | 141 | |
NM_001344167.1 | NP_001332704.1 | 3 - 4 | 783 | 141 | ||
NM_123077.3 | NP_198534.3 | 5 - 6 | 958 | 141 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |