Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CIP4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001344168.1 | NP_001331331.1 | 3 - 4 | 763 | 82 | |
NM_001344169.1 | NP_001331332.1 | 3 - 4 | 720 | 82 | ||
NM_123078.3 | NP_568546.1 | 3 - 4 | 763 | 82 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |