Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SEC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001337509.1 | NP_001326045.1 | 18 - 19 | 2164 | 60 | |
NM_111295.4 | NP_187074.1 | 24 - 25 | 2764 | 60 | ||
GenBank mRNA | AF441079.1 | - | 24 - 25 | 2682 | 60 | |
AK226884.1 | - | 24 - 25 | 2707 | 60 | ||
AY090938.1 | - | 24 - 25 | 2703 | 60 | ||
AY117340.1 | - | 24 - 25 | 2553 | 60 | ||
KJ138841.1 | - | 24 - 25 | 2553 | 60 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |