Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SUS4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001339091.1 | NP_001326640.1 | 1 - 2 | 187 | 89 | |
NM_114187.5 | NP_566865.2 | 2 - 3 | 369 | 89 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |