Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AAO2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_114228.3 | NP_189946.1 | 8 - 9 | 3906 | 121 | |
GenBank mRNA | AB005805.1 | - | 8 - 9 | 3867 | 121 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |