Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GH9A4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001339113.1 | NP_001327805.1 | 5 - 6 | 1100 | 102 | |
NM_114254.3 | NP_189972.2 | 5 - 6 | 1100 | 102 | ||
GenBank mRNA | AY072099.1 | - | 5 - 6 | 1060 | 102 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |