Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CYP71 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_114328.4 | NP_190046.2 | 10 - 11 | 1533 | 88 | |
GenBank mRNA | AY062653.1 | - | 10 - 11 | 1491 | 88 | |
BT008382.1 | - | 10 - 11 | 1430 | 88 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |