Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NUDT5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001335236.1 | NP_001324657.1 | 4 - 5 | 779 | 98 | |
NM_126476.3 | NP_178524.2 | 3 - 4 | 457 | 98 | ||
GenBank mRNA | BT010760.1 | - | 3 - 4 | 457 | 98 | |
BT011326.1 | - | 3 - 4 | 414 | 98 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |