Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MOD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_126612.3 | NP_565331.1 | 3 - 4 | 586 | 74 | |
NM_179609.1 | NP_849940.1 | 3 - 4 | 576 | 74 | ||
GenBank mRNA | AY056192.1 | - | 3 - 4 | 547 | 74 | |
AY113962.1 | - | 2 - 3 | 389 | 74 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |