Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OPR3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001084415.2 | NP_001077884.1 | 2 - 3 | 161 | 93 | |
NM_126619.4 | NP_178662.1 | 2 - 3 | 334 | 93 | ||
NM_201702.2 | NP_973431.1 | 1 - 2 | 282 | 93 | ||
GenBank mRNA | AF370582.1 | - | 1 - 2 | 73 | 93 | |
AF410322.1 | - | 2 - 3 | 118 | 93 | ||
AK317250.1 | - | 1 - 2 | 283 | 93 | ||
AY097367.1 | - | 1 - 2 | 73 | 93 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |