Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ABCC13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001335345.1 | NP_001323939.1 | 21 - 22 | 2268 | 79 | |
NM_001335346.1 | NP_001323940.1 | 21 - 22 | 2268 | 79 | ||
NM_001335347.1 | NP_001323936.1 | 20 - 21 | 2185 | 79 | ||
NM_001335348.1 | NP_001323937.1 | 20 - 21 | 2114 | 79 | ||
NM_001335349.1 | NP_001323938.1 | 18 - 19 | 2045 | 79 | ||
NM_126770.8 | NP_178811.7 | 20 - 21 | 2125 | 79 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |