Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CRCK3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_126855.4 | NP_565351.1 | 5 - 6 | 1279 | 95 | |
GenBank mRNA | AF367339.1 | - | 5 - 6 | 1164 | 95 | |
AY093990.1 | - | 4 - 5 | 936 | 95 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |