Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CRCK3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_126855.4 | NP_565351.1 | 6 - 7 | 1375 | 106 | |
GenBank mRNA | AF367339.1 | - | 6 - 7 | 1260 | 106 | |
AY093990.1 | - | 5 - 6 | 1032 | 106 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |