Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MGDC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001124829.2 | NP_001118301.1 | 1 - 2 | 382 | 78 | |
NM_001335374.1 | NP_001323965.1 | 1 - 2 | 539 | 78 | ||
NM_126865.5 | NP_565352.1 | 1 - 2 | 539 | 78 | ||
GenBank mRNA | AB047398.1 | - | 1 - 2 | 335 | 78 | |
BT026357.1 | - | 1 - 2 | 313 | 78 | ||
KJ138917.1 | - | 1 - 2 | 313 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Arabidopsis thaliana Probe spans exons |