Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MGDC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001124829.2 | NP_001118301.1 | 4 - 5 | 862 | 92 | |
NM_001335374.1 | NP_001323965.1 | 4 - 5 | 1019 | 92 | ||
NM_001335375.1 | NP_001323966.1 | 3 - 4 | 531 | 92 | ||
NM_126865.5 | NP_565352.1 | 4 - 5 | 1019 | 92 | ||
GenBank mRNA | AB047398.1 | - | 4 - 5 | 815 | 92 | |
BT026357.1 | - | 4 - 5 | 793 | 92 | ||
KJ138917.1 | - | 4 - 5 | 793 | 92 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |