Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DRP3B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001161044.1 | NP_001154516.1 | 16 - 17 | 1941 | 98 | |
NM_001335415.1 | NP_001323471.1 | 15 - 16 | 1883 | 98 | ||
NM_126984.4 | NP_565362.1 | 15 - 16 | 2009 | 98 | ||
NM_126985.6 | NP_565363.2 | 15 - 16 | 1854 | 98 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |