Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PDIL5-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001331745.1 | NP_001318951.1 | 2 - 3 | 243 | 126 | |
NM_100670.2 | NP_172274.1 | 2 - 3 | 209 | 126 | ||
NM_202058.2 | NP_973787.1 | 1 - 2 | 601 | 126 | ||
NM_202059.3 | NP_973788.1 | 2 - 3 | 220 | 126 | ||
GenBank mRNA | AK117714.1 | - | 2 - 3 | 210 | 126 | |
BT004994.1 | - | 1 - 2 | 109 | 126 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |