Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SRS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001332005.1 | NP_001321932.1 | 3 - 4 | 540 | 84 | |
NM_001332006.1 | NP_001321931.1 | 3 - 4 | 540 | 84 | ||
NM_001332007.1 | NP_001321930.1 | 1 - 2 | 150 | 84 | ||
NM_101058.2 | NP_172651.1 | 3 - 4 | 485 | 84 | ||
NM_179316.3 | NP_849647.1 | 3 - 4 | 580 | 84 | ||
NM_202083.1 | NP_973812.1 | 3 - 4 | 491 | 84 | ||
GenBank mRNA | AY091115.1 | - | 3 - 4 | 492 | 84 | |
AY122978.1 | - | 3 - 4 | 442 | 84 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |