Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AGL87 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001332553.1 | NP_001319062.1 | 1 | 1579 | 98 | |
GenBank mRNA | AY141250.1 | - | 1 | 161 | 98 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Arabidopsis thaliana Both primers and probe map within a single exon |