Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CSN5A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_102139.2 | NP_173705.1 | 2 - 3 | 831 | 145 | |
NM_202161.1 | NP_973890.1 | 2 - 3 | 821 | 145 | ||
GenBank mRNA | AF042334.1 | - | 2 - 3 | 685 | 145 | |
AF395062.1 | - | 2 - 3 | 665 | 145 | ||
BT029486.1 | - | 2 - 3 | 665 | 145 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |