Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CIB5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001036016.2 | NP_001031093.1 | 8 - 9 | 1063 | 101 | |
NM_001332707.1 | NP_001323071.1 | 8 - 9 | 1300 | 101 | ||
NM_001332709.1 | NP_001323073.1 | 8 - 9 | 1459 | 101 | ||
NM_102390.3 | NP_173950.1 | 7 - 8 | 1566 | 101 | ||
NM_202184.3 | NP_973913.1 | 8 - 9 | 1310 | 101 | ||
GenBank mRNA | AK117355.1 | - | 7 - 8 | 1266 | 101 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |