Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HST | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001337559.1 | NP_001327303.1 | 15 - 16 | 2109 | 144 | |
NM_111376.4 | NP_187155.2 | 15 - 16 | 2121 | 144 | ||
GenBank mRNA | AK229239.1 | - | 15 - 16 | 2016 | 144 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |