Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ALY2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001035566.2 | NP_001030643.1 | 11 - 12 | 1714 | 97 | |
NM_001125108.2 | NP_001118580.1 | 10 - 11 | 1800 | 97 | ||
NM_001202883.1 | NP_001189812.1 | 11 - 12 | 1679 | 97 | ||
NM_001202884.1 | NP_001189813.1 | 12 - 13 | 1647 | 97 | ||
NM_111411.5 | NP_187189.2 | 11 - 12 | 1711 | 97 | ||
GenBank mRNA | AJ583496.1 | - | 11 - 12 | 1651 | 97 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |