Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ACBP4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_111415.5 | NP_187193.3 | 1 - 2 | 255 | 69 | |
NM_202498.2 | NP_974227.1 | 1 - 2 | 362 | 69 | ||
GenBank mRNA | AK228046.1 | - | 1 - 2 | 256 | 69 | |
BT006458.1 | - | 1 - 2 | 153 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |