Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RAD9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001202886.1 | NP_001189815.1 | 7 - 8 | 896 | 92 | |
NM_001337585.1 | NP_001319478.1 | 6 - 7 | 875 | 92 | ||
NM_001337586.1 | NP_001319479.1 | 6 - 7 | 858 | 92 | ||
GenBank mRNA | BT029213.1 | - | 6 - 7 | 764 | 92 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |