Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LON3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_111452.1 | NP_566258.1 | 8 - 9 | 995 | 96 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |