Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LACS6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_111471.3 | NP_566265.1 | 8 - 9 | 793 | 75 | |
GenBank mRNA | AY094418.1 | - | 8 - 9 | 707 | 75 | |
BT001117.1 | - | 8 - 9 | 652 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |