Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PRXIIF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001337629.1 | NP_001327152.1 | 4 - 5 | 813 | 74 | |
NM_111480.4 | NP_566268.1 | 4 - 5 | 602 | 74 | ||
GenBank mRNA | AY052278.1 | - | 4 - 5 | 535 | 74 | |
AY098963.1 | - | 4 - 5 | 505 | 74 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |